Assay Details
| Gene Symbol: MAP1LC3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130558.4 | XP_003130606.1 | 585 | 62 | ||
| XM_013980061.2 | XP_013835515.1 | 393 | 62 | |||
More Information
Set Membership: |
|

MAP1LC3C
Genomic Map