Assay Details
| Gene Symbol: TMEM150B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094999.1 | XP_020950658.1 | 656 | 60 | ||
| XM_021095000.1 | XP_020950659.1 | 653 | 60 | |||
| XM_021095001.1 | XP_020950660.1 | 522 | 60 | |||
| XM_021095002.1 | XP_020950661.1 | 351 | 60 | |||
| XM_021095003.1 | XP_020950662.1 | 661 | 60 | |||
| XM_021095004.1 | XP_020950663.1 | 527 | 60 | |||
More Information
Set Membership: |
|

TMEM150B
Genomic Map