Assay Details
| Gene Symbol: ARHGEF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005657810.3 | XP_005657867.1 | 1445 | 69 | ||
| XM_005657811.3 | XP_005657868.1 | 2148 | 69 | |||
| XM_013990906.2 | XP_013846360.1 | 2160 | 69 | |||
| XM_013990907.2 | XP_013846361.1 | 2156 | 69 | |||
| XM_013990908.2 | XP_013846362.1 | 1975 | 69 | |||
| XM_013990909.2 | XP_013846363.1 | 1979 | 69 | |||
| XM_013990911.2 | XP_013846365.1 | 1968 | 69 | |||
| XM_021080633.1 | XP_020936292.1 | 3598 | 69 | |||
| XM_021080634.1 | XP_020936293.1 | 3358 | 69 | |||
| XM_021080635.1 | XP_020936294.1 | 1316 | 69 | |||
| XM_021080636.1 | XP_020936295.1 | 1231 | 69 | |||
More Information
Set Membership: |
|

ARHGEF9
Genomic Map