Assay Details
| Gene Symbol: LRRN4CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003353820.4 | XP_003353868.2 | 335 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003353820.4 transcripts.
Set Membership: |
|

LRRN4CL
Genomic Map