Assay Details
| Gene Symbol: TMEM45B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021063152.1 | XP_020918811.1 | 416 | 64 | ||
| XM_021063153.1 | XP_020918812.1 | 526 | 64 | |||
| XM_021063154.1 | XP_020918813.1 | 696 | 64 | |||
| XM_021063155.1 | XP_020918814.1 | 339 | 64 | |||
| XM_021063156.1 | XP_020918815.1 | 3111 | 64 | |||
More Information
Set Membership: |
|

TMEM45B
Genomic Map