Assay Details
| Gene Symbol: ANKRD33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091670.1 | XP_020947329.1 | 569 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021091670.1 transcripts.
Set Membership: |
|

ANKRD33
Genomic Map