Assay Details
| Gene Symbol: MRPS33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003360107.4 | XP_003360155.1 | 477 | 85 | ||
| XM_003360108.4 | XP_003360156.1 | 363 | 85 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MRPS33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003360107.4 | XP_003360155.1 | 477 | 85 | ||
| XM_003360108.4 | XP_003360156.1 | 363 | 85 | |||
Set Membership: |
|