Assay Details
| Gene Symbol: NXT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135314.4 | XP_003135362.1 | 468 | 82 | ||
| XM_005673824.2 | XP_005673881.1 | 396 | 82 | |||
| XM_005673825.3 | XP_005673882.1 | 588 | 82 | |||
More Information
Set Membership: |
|

NXT2
Genomic Map