Assay Details
| Gene Symbol: FAM221A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021078732.1 | XP_020934391.1 | 152 | 65 | ||
| XM_021078733.1 | XP_020934392.1 | 152 | 65 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM221A| Gene Symbol: FAM221A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021078732.1 | XP_020934391.1 | 152 | 65 | ||
| XM_021078733.1 | XP_020934392.1 | 152 | 65 | |||
Set Membership: |
|