Assay Details
| Gene Symbol: GCSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097052.1 | XP_020952711.1 | 561 | 108 | ||
| GenBank mRNA | AK392615.1 | - | 4 - 5 | 503 | 108 | |
| AY610147.1 | - | 4 - 5 | 468 | 108 | ||
More Information
Set Membership: |
|

GCSH
Genomic Map