Assay Details
| Gene Symbol: ZFP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013998456.2 | XP_013853910.1 | 2444 | 98 | ||
| XM_013998457.2 | XP_013853911.1 | 2057 | 98 | |||
| XM_013998458.2 | XP_013853912.1 | 1922 | 98 | |||
| XM_013998459.2 | XP_013853913.1 | 1947 | 98 | |||
| XM_013998461.2 | XP_013853915.1 | 2207 | 98 | |||
| XM_021097236.1 | XP_020952895.1 | 1652 | 98 | |||
| XM_021097237.1 | XP_020952896.1 | 2406 | 98 | |||
| XM_021097238.1 | XP_020952897.1 | 2611 | 98 | |||
| XM_021097239.1 | XP_020952898.1 | 2145 | 98 | |||
| XM_021097240.1 | XP_020952899.1 | 1726 | 98 | |||
| XM_021097241.1 | XP_020952900.1 | 1688 | 98 | |||
| XM_021097242.1 | XP_020952901.1 | 2919 | 98 | |||
| GenBank mRNA | AK391314.1 | - | 2 | 1417 | 98 | |
More Information
Set Membership: |
|

ZFP30
Genomic Map