Assay Details
| Gene Symbol: TMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928249.4 | XP_001928284.1 | 2114 | 91 | ||
| XM_021101807.1 | XP_020957466.1 | 2447 | 91 | |||
| GenBank mRNA | AK392555.1 | - | 1 | 622 | 91 | |
More Information
Set Membership: |
|

TMX1
Genomic Map