Assay Details
| Gene Symbol: IL25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005666258.3 | XP_005666315.1 | 3864 | 79 | ||
| GenBank mRNA | EF584511.1 | - | 2 | 323 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: IL25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005666258.3 | XP_005666315.1 | 3864 | 79 | ||
| GenBank mRNA | EF584511.1 | - | 2 | 323 | 79 | |
Set Membership: |
|