Assay Details
Gene Symbol: HOMER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013991326.2 | XP_013846780.2 | 1511 | 98 | ||
XM_021083465.1 | XP_020939124.1 | 1522 | 98 | |||
XM_021083466.1 | XP_020939125.1 | 1604 | 98 | |||
XM_021083467.1 | XP_020939126.1 | 1198 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
