Assay Details
| Gene Symbol: CLDN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161643.1 | NP_001155115.1 | 3 - 4 | 668 | 70 | |
| XM_021085619.1 | XP_020941278.1 | 1303 | 70 | |||
| GenBank mRNA | AK231230.1 | - | 3 - 4 | 668 | 70 | |
| AK391082.1 | - | 3 - 4 | 669 | 70 | ||
| AK400467.1 | - | 3 - 4 | 704 | 70 | ||
| FJ873104.1 | - | 3 - 4 | 668 | 70 | ||
More Information
Set Membership: |
|

CLDN15
Genomic Map