Assay Details
| Gene Symbol: SPESP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177931.1 | NP_001171402.1 | 1 - 2 | 110 | 62 | |
| GenBank mRNA | GU474995.1 | - | 1 - 2 | 110 | 62 | |
More Information
Set Membership: |
|

SPESP1
Genomic Map