Assay Details
| Gene Symbol: nup62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001989.1 | NP_001001989.1 | 12 - 13 | 1599 | 67 | |
| XM_012968168.1 | XP_012823622.1 | 12 - 13 | 1728 | 67 | ||
| XM_018096189.1 | XP_017951678.1 | 11 - 12 | 1575 | 67 | ||
| GenBank mRNA | BC064245.1 | - | 11 - 12 | 1577 | 67 | |
| BC075599.1 | - | 12 - 13 | 1599 | 67 | ||
More Information
Set Membership: |
|

nup62
Genomic Map