Assay Details
| Gene Symbol: eif3m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004794.1 | NP_001004794.1 | 10 - 11 | 1032 | 58 | |
| GenBank mRNA | BC074530.1 | - | 10 - 11 | 1032 | 58 | |
| CR761845.2 | - | 10 - 11 | 1009 | 58 | ||
More Information
Set Membership: |
|

eif3m
Genomic Map