Assay Details
| Gene Symbol: march8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004831.1 | NP_001004831.1 | 5 - 6 | 680 | 75 | |
| XM_012966348.2 | XP_012821802.1 | 7 - 8 | 1595 | 75 | ||
| XM_012966349.2 | XP_012821803.1 | 6 - 7 | 1792 | 75 | ||
| XM_018095334.1 | XP_017950823.1 | 5 - 6 | 1750 | 75 | ||
| GenBank mRNA | BC074623.1 | - | 5 - 6 | 680 | 75 | |
| CR760341.2 | - | 5 - 6 | 644 | 75 | ||
More Information
Set Membership: |
|

march8
Genomic Map