Assay Details
| Gene Symbol: pgam5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004858.1 | NP_001004858.1 | 1 - 2 | 196 | 92 | |
| XM_012959055.2 | XP_012814509.1 | 1 - 2 | 324 | 92 | ||
| GenBank mRNA | BC074682.1 | - | 1 - 2 | 196 | 92 | |
| CT010555.2 | - | 1 - 2 | 184 | 92 | ||
More Information
Set Membership: |
|

pgam5
Genomic Map