Assay Details
| Gene Symbol: ydjc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004876.1 | NP_001004876.1 | 4 - 5 | 669 | 75 | |
| XM_012959278.2 | XP_012814732.1 | 4 - 5 | 572 | 75 | ||
| XM_012959287.2 | XP_012814741.1 | 4 - 5 | 596 | 75 | ||
| XM_012959293.2 | XP_012814747.1 | 4 - 5 | 1375 | 75 | ||
| XM_012959299.2 | XP_012814753.1 | 5 - 6 | 1623 | 75 | ||
| XM_012959307.2 | XP_012814761.1 | 4 - 5 | 592 | 75 | ||
| GenBank mRNA | BC075260.1 | - | 4 - 5 | 669 | 75 | |
More Information
Set Membership: |
|

ydjc
Genomic Map