Assay Details
| Gene Symbol: tdg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004982.1 | NP_001004982.1 | 7 - 8 | 943 | 65 | |
| XM_012958838.2 | XP_012814292.1 | 7 - 8 | 1018 | 65 | ||
| XM_012958839.2 | XP_012814293.1 | 7 - 8 | 894 | 65 | ||
| GenBank mRNA | BC075533.1 | - | 7 - 8 | 943 | 65 | |
| CT030631.1 | - | 7 - 8 | 936 | 65 | ||
More Information
Set Membership: |
|

tdg
Genomic Map