Assay Details
Gene Symbol: poc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004988.1 | NP_001004988.1 | 6 - 7 | 768 | 87 | |
XM_012958840.2 | XP_012814294.1 | 6 - 7 | 886 | 87 | ||
XM_012958841.2 | XP_012814295.1 | 6 - 7 | 888 | 87 | ||
XM_012958842.2 | XP_012814296.1 | 6 - 7 | 885 | 87 | ||
XM_012958844.1 | XP_012814298.1 | 6 - 7 | 740 | 87 | ||
XM_018092031.1 | XP_017947520.1 | 5 - 6 | 663 | 87 | ||
XM_018092032.1 | XP_017947521.1 | 6 - 7 | 885 | 87 | ||
GenBank mRNA | BC075548.1 | - | 6 - 7 | 768 | 87 | |
CU024965.1 | - | 6 - 7 | 819 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
