Assay Details
| Gene Symbol: fcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004999.1 | NP_001004999.1 | 5 - 6 | 439 | 80 | |
| GenBank mRNA | BC075594.1 | - | 5 - 6 | 439 | 80 | |
| CT025502.2 | - | 4 - 5 | 366 | 80 | ||
More Information
Set Membership: |
|

fcf1
Genomic Map