Assay Details
| Gene Symbol: fcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004999.1 | NP_001004999.1 | 7 - 8 | 624 | 70 | |
| GenBank mRNA | BC075594.1 | - | 7 - 8 | 624 | 70 | |
| CT025502.2 | - | 6 - 7 | 551 | 70 | ||
More Information
Set Membership: |
|

fcf1
Genomic Map