Assay Details
Gene Symbol: gsto2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005086.2 | NP_001005086.1 | 5 - 6 | 648 | 80 | |
XM_012966444.2 | XP_012821898.1 | 6 - 7 | 722 | 80 | ||
XM_012966445.1 | XP_012821899.1 | 5 - 6 | 584 | 80 | ||
GenBank mRNA | BC077010.1 | - | 5 - 6 | 649 | 80 | |
CR760625.2 | - | 5 - 6 | 622 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
