Assay Details
Gene Symbol: pcbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011016.2 | NP_001011016.1 | 3 - 4 | 290 | 74 | |
GenBank mRNA | BC082730.1 | - | 3 - 4 | 291 | 74 | |
CR760715.2 | - | 3 - 4 | 232 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
