Assay Details
| Gene Symbol: atp5pd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011021.2 | NP_001011021.1 | 5 - 6 | 429 | 96 | |
| GenBank mRNA | BC082736.1 | - | 5 - 6 | 430 | 96 | |
More Information
Set Membership: |
|

atp5pd
Genomic Map