Assay Details
| Gene Symbol: gcsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011024.1 | NP_001011024.1 | 2 - 3 | 223 | 73 | |
| GenBank mRNA | BC082740.1 | - | 2 - 3 | 223 | 73 | |
| CR761928.2 | - | 2 - 3 | 221 | 73 | ||
More Information
Set Membership: |
|

gcsh
Genomic Map