Assay Details
| Gene Symbol: spred2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011032.1 | NP_001011032.1 | 2 - 3 | 515 | 94 | |
| GenBank mRNA | AY714337.1 | - | 2 - 3 | 515 | 94 | |
| BC171265.1 | - | 2 - 3 | 490 | 94 | ||
| CT027924.3 | - | 2 - 3 | 513 | 94 | ||
More Information
Set Membership: |
|

spred2
Genomic Map