Assay Details
Gene Symbol: sar1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203514.1 | NP_988845.1 | 5 - 6 | 466 | 79 | |
GenBank mRNA | BC063212.1 | - | 5 - 6 | 466 | 79 | |
BC075541.1 | - | 5 - 6 | 530 | 79 | ||
CR762283.2 | - | 5 - 6 | 420 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
