Assay Details
| Gene Symbol: fth1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203677.1 | NP_989008.1 | 3 - 4 | 501 | 66 | |
| GenBank mRNA | BC061303.1 | - | 3 - 4 | 501 | 66 | |
| CT027933.1 | - | 3 - 4 | 445 | 66 | ||
More Information
Set Membership: |
|

fth1
Genomic Map