Assay Details
| Gene Symbol: nosip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203698.1 | NP_989029.1 | 8 - 9 | 930 | 69 | |
| XM_012956642.2 | XP_012812096.1 | 8 - 9 | 1062 | 69 | ||
| GenBank mRNA | BC061402.1 | - | 8 - 9 | 930 | 69 | |
More Information
Set Membership: |
|

nosip
Genomic Map