Assay Details
| Gene Symbol: set | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203710.1 | NP_989041.1 | 5 - 6 | 641 | 104 | |
| XM_012968151.2 | XP_012823605.1 | 4 - 5 | 778 | 104 | ||
| XM_012968152.2 | XP_012823606.1 | 4 - 5 | 781 | 104 | ||
| GenBank mRNA | BC061372.1 | - | 5 - 6 | 641 | 104 | |
| CR762001.2 | - | 4 - 5 | 435 | 104 | ||
More Information
Set Membership: |
|

set
Genomic Map