Assay Details
| Gene Symbol: prdx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203771.1 | NP_989102.1 | 3 - 4 | 408 | 95 | |
| GenBank mRNA | BC062510.1 | - | 3 - 4 | 408 | 95 | |
| BC169178.1 | - | 3 - 4 | 365 | 95 | ||
| CR760860.2 | - | 3 - 4 | 404 | 95 | ||
More Information
Set Membership: |
|

prdx6
Genomic Map