Assay Details
| Gene Symbol: cops5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203778.1 | NP_989109.1 | 1 - 2 | 152 | 62 | |
| GenBank mRNA | BC062499.1 | - | 1 - 2 | 152 | 62 | |
| CR760870.2 | - | 1 - 2 | 145 | 62 | ||
More Information
Set Membership: |
|

cops5
Genomic Map