Assay Details
Gene Symbol: prps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203809.1 | NP_989140.1 | 4 - 5 | 574 | 64 | |
XM_012968153.2 | XP_012823607.1 | 4 - 5 | 434 | 64 | ||
GenBank mRNA | BC061401.1 | - | 4 - 5 | 574 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
