Assay Details
Gene Symbol: lrat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203810.1 | NP_989141.1 | 1 - 2 | 542 | 97 | |
GenBank mRNA | BC061345.1 | - | 1 - 2 | 542 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
