Assay Details
| Gene Symbol: eno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203813.1 | NP_989144.1 | 2 - 3 | 120 | 80 | |
| XM_012966301.1 | XP_012821755.1 | 2 - 3 | 223 | 80 | ||
| XM_012966302.1 | XP_012821756.1 | 2 - 3 | 197 | 80 | ||
| XM_012966304.2 | XP_012821758.1 | 2 - 3 | 193 | 80 | ||
| GenBank mRNA | BC061287.1 | - | 2 - 3 | 120 | 80 | |
| BC158432.1 | - | 2 - 3 | 116 | 80 | ||
| CT025514.2 | - | 2 - 3 | 109 | 80 | ||
More Information
Set Membership: |
|

eno1
Genomic Map