Assay Details
| Gene Symbol: eno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203813.1 | NP_989144.1 | 4 - 5 | 276 | 89 | |
| XM_012966304.2 | XP_012821758.1 | 4 - 5 | 349 | 89 | ||
| GenBank mRNA | BC061287.1 | - | 4 - 5 | 276 | 89 | |
| CT025514.2 | - | 4 - 5 | 265 | 89 | ||
More Information
Set Membership: |
|

eno1
Genomic Map