Assay Details
| Gene Symbol: eno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203813.1 | NP_989144.1 | 7 - 8 | 700 | 65 | |
| XM_012966301.1 | XP_012821755.1 | 7 - 8 | 803 | 65 | ||
| XM_012966302.1 | XP_012821756.1 | 7 - 8 | 777 | 65 | ||
| XM_012966304.2 | XP_012821758.1 | 7 - 8 | 773 | 65 | ||
| GenBank mRNA | BC061287.1 | - | 7 - 8 | 700 | 65 | |
| BC158432.1 | - | 7 - 8 | 696 | 65 | ||
| CT025514.2 | - | 7 - 8 | 689 | 65 | ||
More Information
Set Membership: |
|

eno1
Genomic Map