Assay Details
| Gene Symbol: cpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203862.2 | NP_989193.1 | 3 - 4 | 372 | 69 | |
| XM_012960762.2 | XP_012816216.1 | 4 - 5 | 1390 | 69 | ||
| GenBank mRNA | BC063210.1 | - | 3 - 4 | 375 | 69 | |
| CT027886.1 | - | 3 - 4 | 380 | 69 | ||
More Information
Set Membership: |
|

cpt2
Genomic Map