Assay Details
Gene Symbol: pin4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203870.1 | NP_989201.1 | 3 - 4 | 424 | 73 | |
XM_018096185.1 | XP_017951674.1 | 3 - 4 | 345 | 73 | ||
GenBank mRNA | BC063359.1 | - | 3 - 4 | 424 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
