Assay Details
| Gene Symbol: lypla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203956.1 | NP_989287.1 | 8 - 9 | 752 | 78 | |
| GenBank mRNA | BC064187.1 | - | 8 - 9 | 752 | 78 | |
More Information
Set Membership: |
|

lypla2
Genomic Map