Assay Details
| Gene Symbol: plk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213679.2 | NP_998844.1 | 1 - 2 | 528 | 72 | |
| GenBank mRNA | BC067963.2 | - | 1 - 2 | 528 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

plk1| Gene Symbol: plk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213679.2 | NP_998844.1 | 1 - 2 | 528 | 72 | |
| GenBank mRNA | BC067963.2 | - | 1 - 2 | 528 | 72 | |
Set Membership: |
|