Assay Details
| Gene Symbol: snrpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005128.1 | NP_001005128.1 | 2 - 3 | 255 | 83 | |
| GenBank mRNA | BC077664.1 | - | 2 - 3 | 255 | 83 | |
| BC161312.1 | - | 2 - 3 | 83 | |||
| CR760591.2 | - | 2 - 3 | 83 | |||
More Information
Set Membership: |
|

snrpd2
Genomic Map