Assay Details
| Gene Symbol: ubtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005656.1 | NP_001005656.1 | 1 - 2 | 483 | 71 | |
| XM_012966359.2 | XP_012821813.1 | 1 - 2 | 822 | 71 | ||
| GenBank mRNA | BC074677.1 | - | 1 - 2 | 483 | 71 | |
More Information
Set Membership: |
|

ubtd1
Genomic Map