Assay Details
| Gene Symbol: ubtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005656.1 | NP_001005656.1 | 2 - 3 | 704 | 68 | |
| XM_012966359.2 | XP_012821813.1 | 2 - 3 | 1043 | 68 | ||
| GenBank mRNA | BC074677.1 | - | 2 - 3 | 704 | 68 | |
More Information
Set Membership: |
|

ubtd1
Genomic Map