Assay Details
| Gene Symbol: xkrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006125.1 | NP_001006125.1 | 2 - 3 | 788 | 87 | |
| XM_012968202.2 | XP_012823656.1 | 2 - 3 | 1449 | 87 | ||
| XM_012968203.2 | XP_012823657.1 | 2 - 3 | 1056 | 87 | ||
| XM_012968204.2 | XP_012823658.1 | 2 - 3 | 790 | 87 | ||
| GenBank mRNA | AY534992.1 | - | 1 - 2 | 688 | 87 | |
| BC075344.1 | - | 2 - 3 | 788 | 87 | ||
More Information
Set Membership: |
|

xkrx
Genomic Map