Assay Details
| Gene Symbol: ube2q2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006695.1 | NP_001006696.1 | 3 - 4 | 448 | 71 | |
| GenBank mRNA | BC075400.1 | - | 3 - 4 | 448 | 71 | |
| CR761026.2 | - | 3 - 4 | 422 | 71 | ||
More Information
Set Membership: |
|

ube2q2
Genomic Map